siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107273.1 | NP_001100743.1 | 5 | 578 | ||
| XM_006252117.2 | XP_006252179.1 | 5 | 652 | |||
| XM_006252118.2 | XP_006252180.1 | 5 | 760 | |||
| XM_006252120.2 | XP_006252182.1 | 5 | 652 | |||
| XM_006252121.2 | XP_006252183.1 | 2 | 706 | |||
| XM_008770757.1 | XP_008768979.1 | 2 | 32 | |||
| GenBank mRNA | BC087063.1 | - | 5 | 605 | ||
| BC105847.1 | - | 5 | 648 | |||
| FQ211026.1 | - | 5 | 559 | |||

Cdca2
Genomic Map