siRNA Details
| Gene Symbol: Cdca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107273.1 | NP_001100743.1 | 4 | 488 | ||
| XM_006252117.2 | XP_006252179.1 | 4 | 562 | |||
| XM_006252118.2 | XP_006252180.1 | 4 | 670 | |||
| XM_006252120.2 | XP_006252182.1 | 4 | 562 | |||
| GenBank mRNA | BC087063.1 | - | 4 | 515 | ||
| BC105847.1 | - | 4 | 558 | |||
| FQ211026.1 | - | 4 | 469 | |||

Cdca2
Genomic Map