siRNA Details
| Gene Symbol: Slc25a37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013996.1 | NP_001014018.1 | 4 | 739 | ||
| XM_006252266.1 | XP_006252328.1 | 5 | 911 | |||
| GenBank mRNA | BC082071.1 | - | 4 | 739 | 7132913 | |
| FQ227341.1 | - | 4 | 680 | |||
| FQ230701.1 | - | 3 | 396 | |||
| FQ231671.1 | - | 3 | 210 | |||
| FQ231945.1 | - | 3 | 227 | |||
| FQ233331.1 | - | 4 | 639 | |||

Slc25a37
Genomic Map