siRNA Details
| Gene Symbol: Slc25a37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013996.1 | NP_001014018.1 | 4 | 786 | ||
| XM_006252266.1 | XP_006252328.1 | 5 | 958 | |||
| GenBank mRNA | BC082071.1 | - | 4 | 786 | 7132913 | |
| FQ227341.1 | - | 4 | 727 | |||
| FQ227813.1 | - | 1 | 121 | |||
| FQ231671.1 | - | 3 | 257 | |||
| FQ231945.1 | - | 3 | 274 | |||
| FQ233331.1 | - | 4 | 686 | |||

Slc25a37
Genomic Map