siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170472.1 | NP_001163943.1 | 19 | 2618 | ||
| XM_006252268.2 | XP_006252330.1 | 19 | 2735 | |||
| GenBank mRNA | BC169115.1 | - | 5 | 631 | ||
| CO557150.1 | - | 2 | 235 | |||
| EV777431.1 | - | 6 | 717 | |||
| FQ211644.1 | - | 19 | 2729 | |||

Ccar2
Genomic Map