siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170472.1 | NP_001163943.1 | 4 | 306 | ||
| XM_006252268.2 | XP_006252330.1 | 4 | 423 | |||
| GenBank mRNA | CK596250.1 | - | 1 | 55 | ||
| FM034041.1 | - | 4 | 306 | |||
| FQ211644.1 | - | 4 | 417 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_146055, BC038346, AK050238, AK028194, BC021475.

Ccar2
Genomic Map