siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170472.1 | NP_001163943.1 | 14 | 1853 | ||
| XM_006252268.2 | XP_006252330.1 | 14 | 1970 | |||
| GenBank mRNA | FQ211644.1 | - | 14 | 1964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccar2| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170472.1 | NP_001163943.1 | 14 | 1853 | ||
| XM_006252268.2 | XP_006252330.1 | 14 | 1970 | |||
| GenBank mRNA | FQ211644.1 | - | 14 | 1964 | ||