siRNA Details
| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107275.1 | NP_001100745.1 | 7 | 990 | ||
| XM_006252270.2 | XP_006252332.1 | 7 | 1015 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a14| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107275.1 | NP_001100745.1 | 7 | 990 | ||
| XM_006252270.2 | XP_006252332.1 | 7 | 1015 | |||