siRNA Details
| Gene Symbol: Piwil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107276.1 | NP_001100746.1 | 16 | 2124 | ||
| XM_006252271.2 | XP_006252333.2 | 16 | 2070 | |||
| XM_008770828.1 | XP_008769050.1 | 15 | 1960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piwil2| Gene Symbol: Piwil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107276.1 | NP_001100746.1 | 16 | 2124 | ||
| XM_006252271.2 | XP_006252333.2 | 16 | 2070 | |||
| XM_008770828.1 | XP_008769050.1 | 15 | 1960 | |||