siRNA Details
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170474.1 | NP_001163945.1 | 5, 6 | 563 | ||
XM_006252274.2 | XP_006252336.1 | 5, 6 | 1060 | |||
GenBank mRNA | BC168691.1 | - | 5, 6 | 563 | 7934515 |
