siRNA Details
| Gene Symbol: Fam160b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170474.1 | NP_001163945.1 | 14, 15 | 1891 | ||
| XM_006252274.2 | XP_006252336.1 | 14, 15 | 2388 | |||
| GenBank mRNA | BC168691.1 | - | 14, 15 | 1891 | 7934515 | |

Fam160b2
Genomic Map