siRNA Details
| Gene Symbol: Bora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013997.1 | NP_001014019.1 | 10 | 1243 | ||
| XM_008770868.1 | XP_008769090.1 | 10 | 1601 | |||
| GenBank mRNA | BC088204.1 | - | 10 | 1243 | 7380234 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bora| Gene Symbol: Bora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013997.1 | NP_001014019.1 | 10 | 1243 | ||
| XM_008770868.1 | XP_008769090.1 | 10 | 1601 | |||
| GenBank mRNA | BC088204.1 | - | 10 | 1243 | 7380234 | |