siRNA Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127483.1 | NP_001120955.1 | 16 | 2141 | ||
| XM_006252391.2 | XP_006252453.1 | 17 | 2002 | |||
| XM_008770869.1 | XP_008769091.1 | 16 | 2076 | |||
| GenBank mRNA | BC166824.1 | - | 16 | 2141 | 8370691 | |
| FQ210067.1 | - | 16 | 2153 | |||

Dis3
Genomic Map