siRNA Details
| Gene Symbol: Fbxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100568.1 | NP_001094038.1 | 2 | 386 | ||
| XM_006252442.2 | XP_006252504.1 | 2 | 603 | |||
| GenBank mRNA | CB718262.1 | - | 2 | 386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl3| Gene Symbol: Fbxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100568.1 | NP_001094038.1 | 2 | 386 | ||
| XM_006252442.2 | XP_006252504.1 | 2 | 603 | |||
| GenBank mRNA | CB718262.1 | - | 2 | 386 | ||