siRNA Details
| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005554.1 | NP_001005554.1 | 6 | 815 | ||
| GenBank mRNA | BC081873.1 | - | 6 | 815 | 7107003 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm9sf2| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005554.1 | NP_001005554.1 | 6 | 815 | ||
| GenBank mRNA | BC081873.1 | - | 6 | 815 | 7107003 | |