siRNA Details
| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100685.1 | NP_001094155.1 | 2, 3 | 269 | ||
| GenBank mRNA | BC088243.1 | - | 2, 3 | 249 | ||
| FM063772.1 | - | 2, 3 | 269 | |||
| FQ214276.1 | - | 1, 2 | 54 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clybl| Gene Symbol: Clybl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100685.1 | NP_001094155.1 | 2, 3 | 269 | ||
| GenBank mRNA | BC088243.1 | - | 2, 3 | 249 | ||
| FM063772.1 | - | 2, 3 | 269 | |||
| FQ214276.1 | - | 1, 2 | 54 | |||