siRNA Details
| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107289.1 | NP_001100759.1 | 3, 4 | 219 | ||
| XM_008762881.1 | XP_008761103.1 | 3, 4 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm5| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107289.1 | NP_001100759.1 | 3, 4 | 219 | ||
| XM_008762881.1 | XP_008761103.1 | 3, 4 | 559 | |||