siRNA Details
| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107289.1 | NP_001100759.1 | 2 | 146 | ||
| XM_008762881.1 | XP_008761103.1 | 2 | 486 | |||
| GenBank mRNA | FQ224115.1 | - | 2 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm5| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107289.1 | NP_001100759.1 | 2 | 146 | ||
| XM_008762881.1 | XP_008761103.1 | 2 | 486 | |||
| GenBank mRNA | FQ224115.1 | - | 2 | 110 | ||