siRNA Details
| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013998.2 | NP_001014020.2 | 3 | 1578 | ||
| GenBank mRNA | AY387060.1 | - | 3 | 1578 | ||
| BC079396.1 | - | 3 | 1762 | 7112419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde12| Gene Symbol: Pde12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013998.2 | NP_001014020.2 | 3 | 1578 | ||
| GenBank mRNA | AY387060.1 | - | 3 | 1578 | ||
| BC079396.1 | - | 3 | 1762 | 7112419 | ||