siRNA Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771024.1 | XP_008769246.1 | 4 | 420 | ||
| XM_008771025.1 | XP_008769247.1 | 4 | 420 | |||
| XM_008771026.1 | XP_008769248.1 | 5 | 504 | |||
| XM_008771027.1 | XP_008769249.1 | 4 | 478 | |||
| XM_008771028.1 | XP_008769250.1 | 4 | 420 | |||
| XM_008771029.1 | XP_008769251.1 | 4 | 420 | |||

Ccdc66
Genomic Map