siRNA Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107290.1 | NP_001100760.1 | 6 | 596 | ||
| XM_006252603.2 | XP_006252665.1 | 6 | 578 | |||
| GenBank mRNA | BC168886.1 | - | 2 | 141 | 7369142 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17rb| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107290.1 | NP_001100760.1 | 6 | 596 | ||
| XM_006252603.2 | XP_006252665.1 | 6 | 578 | |||
| GenBank mRNA | BC168886.1 | - | 2 | 141 | 7369142 | |