siRNA Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013134.2 | NP_001013152.2 | 7 | 1240 | ||
| XM_008770996.1 | XP_008769218.1 | 7 | 1344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek4| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013134.2 | NP_001013152.2 | 7 | 1240 | ||
| XM_008770996.1 | XP_008769218.1 | 7 | 1344 | |||