siRNA Details
| Gene Symbol: Glt8d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007683.1 | NP_001007684.1 | 6, 7 | 718 | ||
| GenBank mRNA | AY325256.1 | - | 6, 7 | 665 | ||
| BC079066.1 | - | 6, 7 | 718 | 7121464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glt8d1| Gene Symbol: Glt8d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007683.1 | NP_001007684.1 | 6, 7 | 718 | ||
| GenBank mRNA | AY325256.1 | - | 6, 7 | 665 | ||
| BC079066.1 | - | 6, 7 | 718 | 7121464 | ||