siRNA Details
| Gene Symbol: Ercc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107296.1 | NP_001100766.1 | 10 | 1924 | ||
| XM_006252734.2 | XP_006252796.1 | 11 | 2310 | |||
| XM_006252735.2 | XP_006252797.1 | 11 | 2252 | |||
| XM_006252736.2 | XP_006252798.1 | 11 | 2459 | |||
| XM_006252737.2 | XP_006252799.1 | 10 | 2151 | |||
| XM_006252738.1 | XP_006252800.1 | 8 | 1617 | |||

Ercc6
Genomic Map