siRNA Details
Gene Symbol: USP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004651.3 | NP_004642.2 | 4 | 581 | ||
XM_005272674.3 | XP_005272731.1 | 4 | 418 | |||
XM_011543988.1 | XP_011542290.1 | 4 | 375 | |||
GenBank mRNA | AB073597.1 | - | 4 | 483 | ||
AK055588.1 | - | 3 | 638 | |||
AK098394.1 | - | 4 | 485 | |||
AK294635.1 | - | 4 | 327 | |||
BC000350.2 | - | 4 | 455 | |||
BC063668.1 | - | 4 | 463 | |||
BC140849.1 | - | 4 | 630 | 9021717 | ||
U44839.1 | - | 4 | 441 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
