siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107299.1 | NP_001100769.1 | 6 | 763 | ||
| XM_008771101.1 | XP_008769323.1 | 6 | 751 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgr| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107299.1 | NP_001100769.1 | 6 | 763 | ||
| XM_008771101.1 | XP_008769323.1 | 6 | 751 | |||
Set Membership: |
|