siRNA Details
Gene Symbol: LOC100363321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252792.1 | XP_006252854.1 | 1 | 1034 |
Gene Symbol: LOC100360061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729626.3 | XP_002729672.1 | 1022 |
Gene Symbol: LOC100909786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750168.3 | XP_003750216.1 | 1 | 1022 |
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 1103 |
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013135.2 | NP_001013153.2 | 9 | 1269 | ||
XM_006233681.2 | XP_006233743.1 | 8 | 1179 | |||
XM_006233682.2 | XP_006233744.1 | 8 | 1098 | |||
XM_006233683.2 | XP_006233745.1 | 9 | 1158 | |||
XM_006233684.2 | XP_006233746.1 | 7 | 1011 | |||
XM_006233685.2 | XP_006233747.1 | 7 | 1035 | |||
XM_006233686.2 | XP_006233748.1 | 6 | 999 | |||
GenBank mRNA | BC088296.1 | - | 3 | 209 | 7322728 | |
BC168170.1 | - | 4 | 2556 | 9025284 |
