siRNA Details
| Gene Symbol: Ap1m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044239.1 | NP_001037704.1 | 8 | 918 | ||
| GenBank mRNA | BC107903.1 | - | 8 | 918 | 7455976 | |
| FQ212761.1 | - | 2 | 79 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1m1| Gene Symbol: Ap1m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044239.1 | NP_001037704.1 | 8 | 918 | ||
| GenBank mRNA | BC107903.1 | - | 8 | 918 | 7455976 | |
| FQ212761.1 | - | 2 | 79 | |||