siRNA Details
| Gene Symbol: Ap1m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044239.1 | NP_001037704.1 | 7, 8 | 888 | ||
| GenBank mRNA | BC107903.1 | - | 7, 8 | 888 | 7455976 | |
| FQ212761.1 | - | 1, 2 | 49 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap1m1| Gene Symbol: Ap1m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044239.1 | NP_001037704.1 | 7, 8 | 888 | ||
| GenBank mRNA | BC107903.1 | - | 7, 8 | 888 | 7455976 | |
| FQ212761.1 | - | 1, 2 | 49 | |||