siRNA Details
| Gene Symbol: Ell | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107304.1 | NP_001100774.1 | 11 | 1690 | ||
| XM_008771106.1 | XP_008769328.1 | 11 | 1577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ell| Gene Symbol: Ell | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107304.1 | NP_001100774.1 | 11 | 1690 | ||
| XM_008771106.1 | XP_008769328.1 | 11 | 1577 | |||