siRNA Details
| Gene Symbol: Tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271055.1 | NP_001257984.1 | 3 | 369 | ||
| XM_006252897.2 | XP_006252959.1 | 4 | 439 | |||
| GenBank mRNA | BC089056.1 | - | 3 | 311 | ||
| FQ212329.1 | - | 3 | 343 | |||
| FQ214027.1 | - | 3 | 323 | |||

Tmem59l
Genomic Map