siRNA Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107308.1 | NP_001100778.1 | 9 | 780 | ||
| XM_006252903.2 | XP_006252965.1 | 9 | 1230 | |||
| XM_008771109.1 | XP_008769331.1 | 9 | 1230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmip| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107308.1 | NP_001100778.1 | 9 | 780 | ||
| XM_006252903.2 | XP_006252965.1 | 9 | 1230 | |||
| XM_008771109.1 | XP_008769331.1 | 9 | 1230 | |||