siRNA Details
| Gene Symbol: Sh2d4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012048.1 | NP_001012048.1 | 3 | 482 | ||
| XM_006253032.2 | XP_006253094.1 | 3 | 495 | |||
| GenBank mRNA | BC079100.1 | - | 3 | 482 | 7124466 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d4a| Gene Symbol: Sh2d4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012048.1 | NP_001012048.1 | 3 | 482 | ||
| XM_006253032.2 | XP_006253094.1 | 3 | 495 | |||
| GenBank mRNA | BC079100.1 | - | 3 | 482 | 7124466 | |