siRNA Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253039.2 | XP_006253101.1 | 9 | 3310 | ||
| XM_006253047.2 | XP_006253109.1 | 4 | 1261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psd3| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253039.2 | XP_006253101.1 | 9 | 3310 | ||
| XM_006253047.2 | XP_006253109.1 | 4 | 1261 | |||