siRNA Details
| Gene Symbol: Naf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024772.1 | NP_001019943.1 | 3 | 596 | ||
| GenBank mRNA | BC094302.1 | - | 3 | 596 | 7311858 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naf1| Gene Symbol: Naf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024772.1 | NP_001019943.1 | 3 | 596 | ||
| GenBank mRNA | BC094302.1 | - | 3 | 596 | 7311858 | |