siRNA Details
| Gene Symbol: Ddx60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771220.1 | XP_008769442.1 | 34 | 4653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx60| Gene Symbol: Ddx60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771220.1 | XP_008769442.1 | 34 | 4653 | ||