siRNA Details
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198764.1 | NP_942059.1 | 2 | 498 | ||
XM_008771225.1 | XP_008769447.1 | 1 | 615 | |||
GenBank mRNA | AF515735.1 | - | 2 | 498 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198764.1 | NP_942059.1 | 2 | 498 | ||
XM_008771225.1 | XP_008769447.1 | 1 | 615 | |||
GenBank mRNA | AF515735.1 | - | 2 | 498 |