siRNA Details
| Gene Symbol: Mtus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178093.2 | NP_835194.2 | 9 | 1265 | ||
| XM_006253180.2 | XP_006253242.1 | 13 | 3880 | |||
| XM_006253181.2 | XP_006253243.1 | 13 | 3580 | |||
| XM_006253182.2 | XP_006253244.1 | 11 | 2200 | |||
| XM_008771280.1 | XP_008769502.1 | 10 | 1624 | |||
| GenBank mRNA | AB190507.1 | - | 9 | 1342 | ||
| BC072537.1 | - | 9 | 1265 | |||
| EU417559.1 | - | 12 | 3383 | |||
| GU245886.1 | - | 11 | 1273 | |||

Mtus1
Genomic Map