siRNA Details
| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107312.1 | NP_001100782.1 | 10 | 1175 | ||
| XM_006253195.2 | XP_006253257.1 | 11 | 1314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr7| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107312.1 | NP_001100782.1 | 10 | 1175 | ||
| XM_006253195.2 | XP_006253257.1 | 11 | 1314 | |||