siRNA Details
| Gene Symbol: Dusp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012352.1 | NP_001012352.1 | 2 | 494 | ||
| XM_006253232.2 | XP_006253294.1 | 2 | 342 | |||
| GenBank mRNA | BC089954.1 | - | 2 | 494 | 7312637 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp26| Gene Symbol: Dusp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012352.1 | NP_001012352.1 | 2 | 494 | ||
| XM_006253232.2 | XP_006253294.1 | 2 | 342 | |||
| GenBank mRNA | BC089954.1 | - | 2 | 494 | 7312637 | |