siRNA Details
| Gene Symbol: Htra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107321.1 | NP_001100791.1 | 4 | 1083 | ||
| XM_008771355.1 | XP_008769577.1 | 4 | 4247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htra4| Gene Symbol: Htra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107321.1 | NP_001100791.1 | 4 | 1083 | ||
| XM_008771355.1 | XP_008769577.1 | 4 | 4247 | |||