siRNA Details
| Gene Symbol: Ank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107322.2 | NP_001100792.1 | 32 | 3893 | ||
| XM_008771356.1 | XP_008769578.1 | 35 | 4374 | |||
| XM_008771357.1 | XP_008769579.1 | 34 | 4349 | |||
| XM_008771358.1 | XP_008769580.1 | 35 | 5514 | |||
| XM_008771359.1 | XP_008769581.1 | 35 | 4341 | |||

Ank1
Genomic Map