siRNA Details
Gene Symbol: TMEM187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003492.2 | NP_003483.1 | 2 | 705 | ||
XM_011531199.2 | XP_011529501.1 | 2 | 720 | |||
GenBank mRNA | AK314938.1 | - | 2 | 485 | ||
BC008203.1 | - | 2 | 453 | 3868838 | ||
BC017026.1 | - | 2 | 440 | 3847234 | ||
CR457047.1 | - | 1 | 171 | |||
DQ655925.2 | - | 3 | 438 | |||
X92475.1 | - | 2 | 455 |
