siRNA Details
| Gene Symbol: Myom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169141.1 | NP_001162612.1 | 26 | 3362 | ||
| XM_006253389.2 | XP_006253451.1 | 26 | 3463 | |||
| XM_008771360.1 | XP_008769582.1 | 26 | 3280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myom2| Gene Symbol: Myom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169141.1 | NP_001162612.1 | 26 | 3362 | ||
| XM_006253389.2 | XP_006253451.1 | 26 | 3463 | |||
| XM_008771360.1 | XP_008769582.1 | 26 | 3280 | |||