siRNA Details
| Gene Symbol: Kbtbd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107326.1 | NP_001100796.1 | 1 | 2246 | ||
| XM_006253390.2 | XP_006253452.1 | 2 | 2976 | |||
| XM_006253391.2 | XP_006253453.1 | 2 | 2971 | |||
| XM_006253392.2 | XP_006253454.1 | 2 | 2814 | |||
| XM_006253393.2 | XP_006253455.1 | 2 | 2854 | |||

Kbtbd11
Genomic Map