siRNA Details
| Gene Symbol: Arhgef10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728436.4 | XP_002728482.2 | 16 | 2033 | ||
| XM_006253400.2 | XP_006253462.1 | 16 | 2367 | |||
| XM_006253401.2 | XP_006253463.1 | 16 | 3059 | |||
| XM_006253402.2 | XP_006253464.1 | 17 | 1982 | |||
| XM_006253403.2 | XP_006253465.1 | 15 | 1925 | |||
| XM_008771372.1 | XP_008769594.1 | 16 | 2033 | |||

Arhgef10
Genomic Map