siRNA Details
| Gene Symbol: Arhgef10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728436.4 | XP_002728482.2 | 4 | 708 | ||
| XM_006253400.2 | XP_006253462.1 | 4 | 1042 | |||
| XM_006253401.2 | XP_006253463.1 | 4 | 1734 | |||
| XM_006253402.2 | XP_006253464.1 | 5 | 657 | |||
| XM_006253403.2 | XP_006253465.1 | 4 | 708 | |||
| XM_008771372.1 | XP_008769594.1 | 4 | 708 | |||

Arhgef10
Genomic Map