siRNA Details
| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107329.1 | NP_001100799.1 | 2 | 1929 | ||
| XM_006253396.1 | XP_006253458.1 | 2 | 1920 | |||
| GenBank mRNA | BC168662.1 | - | 2 | 1926 | 7930141 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Champ1| Gene Symbol: Champ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107329.1 | NP_001100799.1 | 2 | 1929 | ||
| XM_006253396.1 | XP_006253458.1 | 2 | 1920 | |||
| GenBank mRNA | BC168662.1 | - | 2 | 1926 | 7930141 | |