siRNA Details
| Gene Symbol: Dapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107335.2 | NP_001100805.2 | 5 | 749 | ||
| XM_006253522.2 | XP_006253584.1 | 4 | 592 | |||
| XM_006253523.2 | XP_006253585.1 | 4 | 592 | |||
| XM_008771440.1 | XP_008769662.1 | 4 | 592 | |||
| XM_008771441.1 | XP_008769663.1 | 4 | 592 | |||
More Information
Set Membership: |
|

Dapk1
Genomic Map