siRNA Details
| Gene Symbol: Spata31d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771468.1 | XP_008769690.1 | 4 | 1424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata31d3| Gene Symbol: Spata31d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771468.1 | XP_008769690.1 | 4 | 1424 | ||